siRNA Details
| Gene Symbol: GID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024052.4 | NP_076957.3 | 6 | 1022 | ||
| XM_005256800.1 | XP_005256857.1 | 5 | 778 | |||
| GenBank mRNA | AK074284.1 | - | 6 | 897 | ||
| AK127580.1 | - | 6 | 709 | |||
| AK315083.1 | - | 6 | 910 | |||
| BC000636.2 | - | 6 | 683 | |||
| BC041829.1 | - | 6 | 1027 | 5268071 | ||

GID4
Genomic Map