siRNA Details
| Gene Symbol: GID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024052.4 | NP_076957.3 | 3 | 726 | ||
| XM_005256799.4 | XP_005256856.1 | 3 | 613 | |||
| XM_005256800.1 | XP_005256857.1 | 3 | 613 | |||
| XM_011524008.1 | XP_011522310.1 | 3 | 613 | |||
| GenBank mRNA | AK074284.1 | - | 3 | 601 | ||
| AK127580.1 | - | 3 | 413 | |||
| AK315083.1 | - | 3 | 614 | |||
| BC000636.2 | - | 3 | 387 | |||
| BC041829.1 | - | 3 | 731 | 5268071 | ||

GID4
Genomic Map