siRNA Details
| Gene Symbol: NUP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024057.3 | NP_076962.2 | 8, 9 | 928 | ||
| GenBank mRNA | AF514994.1 | - | 7, 8 | 782 | ||
| AK026271.1 | - | 7, 8 | 763 | |||
| BC000861.2 | - | 7, 8 | 846 | 3459639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUP37| Gene Symbol: NUP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024057.3 | NP_076962.2 | 8, 9 | 928 | ||
| GenBank mRNA | AF514994.1 | - | 7, 8 | 782 | ||
| AK026271.1 | - | 7, 8 | 763 | |||
| BC000861.2 | - | 7, 8 | 846 | 3459639 | ||