siRNA Details
| Gene Symbol: ZFYVE21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198953.1 | NP_001185882.1 | 4 | 514 | ||
| NM_024071.3 | NP_076976.1 | 4 | 514 | |||
| GenBank mRNA | AK057816.1 | - | 4 | 435 | ||
| AK226142.1 | - | 2 | 3477 | |||
| AK290519.1 | - | 4 | 427 | |||
| AY395756.1 | - | 4 | 429 | |||
| BC001130.2 | - | 4 | 435 | 2966683 | ||
| BC005999.1 | - | 4 | 450 | 4091809 | ||
| BX248261.1 | - | 4 | 420 | |||
| CX786145.1 | - | 4 | 514 | |||

ZFYVE21
Genomic Map