siRNA Details
| Gene Symbol: CRYBA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005208.4 | NP_005199.2 | 3 | 120 | ||
| XM_017024198.1 | XP_016879687.1 | 4 | 416 | |||
| GenBank mRNA | BC069537.1 | - | 3 | 113 | 7262224 | |
| BC113693.1 | - | 3 | 114 | 8322745 | ||
| KM230602.1 | - | 3 | 102 | |||
| KM230603.1 | - | 3 | 101 | |||
| KM230604.1 | - | 3 | 98 | |||
| KM230605.1 | - | 2 | 98 | |||
| KR336660.1 | - | 3 | 98 | |||

CRYBA1
Genomic Map