siRNA Details
| Gene Symbol: NOC4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024078.2 | NP_076983.1 | 11 | 1080 | ||
| XM_017019960.1 | XP_016875449.1 | 10 | 1034 | |||
| GenBank mRNA | BC001191.1 | - | 11 | 1059 | 3354945 | |
| BC007893.2 | - | 11 | 952 | |||
| BC071958.1 | - | 11 | 1023 | 6597539 | ||

NOC4L
Genomic Map