siRNA Details
| Gene Symbol: TMEM109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024092.2 | NP_076997.1 | 4 | 1584 | ||
| GenBank mRNA | AF131803.1 | - | 1 | 429 | ||
| AX775849.1 | - | 5 | 1180 | |||
| AX775851.1 | - | 4 | 1278 | |||
| BC001309.1 | - | 4 | 1278 | 3453616 | ||

TMEM109
Genomic Map