siRNA Details
| Gene Symbol: C2orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286537.1 | NP_001273466.1 | 2 | 426 | ||
| NM_024093.2 | NP_076998.1 | 2 | 426 | |||
| XM_017004892.1 | XP_016860381.1 | 2 | 528 | |||
| GenBank mRNA | AK127661.1 | - | 2 | 231 | ||
| AK304268.1 | - | 2 | 426 | |||
| AK307891.1 | - | 2 | 253 | |||
| BC001310.1 | - | 2 | 229 | 3453623 | ||

C2orf49
Genomic Map