siRNA Details
| Gene Symbol: CCDC86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024098.3 | NP_077003.1 | 1 | 869 | ||
| GenBank mRNA | AK025974.1 | - | 1 | 755 | ||
| AK123775.1 | - | 3 | 542 | |||
| AK302329.1 | - | 2 | 659 | |||
| BC001378.1 | - | 1 | 720 | 3051044 | ||
| DQ501251.1 | - | 1 | 700 | |||

CCDC86
Genomic Map