siRNA Details
| Gene Symbol: CRYBB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001887.3 | NP_001878.1 | 5 | 552 | ||
| XM_011529899.2 | XP_011528201.1 | 5 | 618 | |||
| GenBank mRNA | BC036790.1 | - | 5 | 599 | 5752689 | |
| CR456425.1 | - | 5 | 521 | |||
| U35340.1 | - | 5 | 554 | |||

CRYBB1
Genomic Map