siRNA Details
| Gene Symbol: SMIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300925.1 | NP_001287854.1 | 4 | 209 | ||
| NM_024104.3 | NP_077009.2 | 4 | 209 | |||
| GenBank mRNA | AK025602.1 | - | 4 | 166 | ||
| AL702263.1 | - | 4 | 210 | |||
| BC069279.1 | - | 4 | 182 | |||
| DB047166.1 | - | 4 | 194 | |||

SMIM7
Genomic Map