siRNA Details
| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270891.1 | NP_001257820.1 | 6 | 636 | ||
| NM_001270892.1 | NP_001257821.1 | 5 | 610 | |||
| NM_001270893.1 | NP_001257822.1 | 5 | 568 | |||
| NM_024108.2 | NP_077013.1 | 6 | 678 | |||
| XM_017027295.1 | XP_016882784.1 | 6 | 774 | |||
| GenBank mRNA | AF161407.1 | - | 6 | 630 | ||
| BC001907.1 | - | 6 | 662 | 3534111 | ||
| BC004450.1 | - | 6 | 663 | 2820830 | ||

TRAPPC6A
Genomic Map