siRNA Details
| Gene Symbol: C9orf16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024112.3 | NP_077017.1 | 2 | 683 | ||
| XM_011519004.1 | XP_011517306.1 | 2 | 631 | |||
| GenBank mRNA | AK022885.1 | - | 1 | 2832 | ||
| BC001857.2 | - | 2 | 587 | 3529116 | ||
| BC008887.2 | - | 2 | 587 | 3027807 | ||
| CN360246.1 | - | 2 | 483 | |||

C9orf16
Genomic Map