siRNA Details
| Gene Symbol: TRIM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024114.3 | NP_077019.2 | 2 | 336 | ||
| GenBank mRNA | AF521869.1 | - | 2 | 343 | ||
| BC001862.2 | - | 2 | 336 | 3603763 | ||
| Gene Symbol: TRIM49D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206627.1 | NP_001193556.1 | 1 | 202 | ||
| XM_011542816.2 | XP_011541118.1 | 2 | 318 | |||
| Gene Symbol: TRIM49D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105522.1 | NP_001098992.1 | 2 | 316 | ||

TRIM48
Genomic Map