siRNA Details
| Gene Symbol: PLEKHF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024310.4 | NP_077286.3 | 2 | 1597 | ||
| XM_005259256.3 | XP_005259313.1 | 2 | 1885 | |||
| XM_011527309.2 | XP_011525611.1 | 2 | 2871 | |||
| GenBank mRNA | AK027758.1 | - | 2 | 1556 | ||
| AY037145.2 | - | 2 | 1962 | |||
| BC002744.2 | - | 2 | 1551 | 3628796 | ||
| BC018876.2 | - | 2 | 1551 | |||

PLEKHF1
Genomic Map