siRNA Details
| Gene Symbol: LENG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024316.2 | NP_077292.2 | 1, 2 | 182 | ||
| GenBank mRNA | BC014986.1 | - | 1, 2 | 147 | 4877885 | |
| BP870984.1 | - | 1, 2 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LENG1| Gene Symbol: LENG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024316.2 | NP_077292.2 | 1, 2 | 182 | ||
| GenBank mRNA | BC014986.1 | - | 1, 2 | 147 | 4877885 | |
| BP870984.1 | - | 1, 2 | 183 | |||