siRNA Details
| Gene Symbol: C1orf35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024319.3 | NP_077295.1 | 8 | 1399 | ||
| NR_130975.1 | 9 | 1579 | ||||
| GenBank mRNA | AK123377.1 | - | 5 | 2224 | ||
| AK126087.1 | - | 9 | 3528 | |||
| AY137773.1 | - | 8 | 1260 | |||
| BC002843.2 | - | 8 | 1258 | 3634226 | ||
| BC062585.1 | - | 7 | 1399 | 5212995 | ||

C1orf35
Genomic Map