siRNA Details
| Gene Symbol: C19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024323.3 | NP_077299.3 | 6 | 1071 | ||
| XM_006722885.3 | XP_006722948.2 | 6 | 1444 | |||
| XM_011528286.2 | XP_011526588.1 | 5 | 1233 | |||
| XM_011528287.2 | XP_011526589.1 | 5 | 893 | |||
| XM_017027300.1 | XP_016882789.1 | 5 | 1231 | |||
| XM_017027301.1 | XP_016882790.1 | 5 | 1095 | |||
| GenBank mRNA | AK097431.1 | - | 6 | 1071 | ||
| BC002891.2 | - | 1 | 197 | |||
| BC012945.1 | - | 2 | 647 | |||
| BC119719.1 | - | 5 | 1039 | 40117748 | ||

C19orf57
Genomic Map