siRNA Details
| Gene Symbol: THTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126339.3 | NP_001119811.1 | 2 | 896 | ||
| NM_024328.5 | NP_077304.1 | 1 | 1295 | |||
| GenBank mRNA | AF432862.1 | - | 1 | 564 | ||
| AK057691.1 | - | 1 | 1002 | |||
| AK098604.1 | - | 2 | 685 | |||
| BC002984.1 | - | 1 | 989 | 3535451 | ||
| BI522644.1 | - | 2 | 700 | |||
| BX161435.1 | - | 1 | 1255 | |||

THTPA
Genomic Map