siRNA Details
| Gene Symbol: IRX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024337.3 | NP_077313.3 | 4 | 1816 | ||
| XM_017009845.1 | XP_016865334.1 | 3 | 1942 | |||
| GenBank mRNA | AY335938.1 | - | 4 | 1819 | ||
| BC029160.1 | - | 4 | 1518 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRX1| Gene Symbol: IRX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024337.3 | NP_077313.3 | 4 | 1816 | ||
| XM_017009845.1 | XP_016865334.1 | 3 | 1942 | |||
| GenBank mRNA | AY335938.1 | - | 4 | 1819 | ||
| BC029160.1 | - | 4 | 1518 | |||