siRNA Details
| Gene Symbol: THOC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142350.1 | NP_001135822.1 | 7 | 743 | ||
| NM_024339.3 | NP_077315.2 | 7 | 743 | |||
| GenBank mRNA | AK075330.1 | - | 7 | 710 | ||
| AK314086.1 | - | 7 | 721 | |||
| AL832548.1 | - | 6 | 347 | |||
| BC003118.1 | - | 8 | 623 | 3537243 | ||
| BC050674.1 | - | 7 | 682 | 6067702 | ||

THOC6
Genomic Map