siRNA Details
| Gene Symbol: OR5H6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005479.1 | NP_001005479.1 | 1 | 236 | ||
| XM_011513118.2 | XP_011511420.1 | 2 | 282 | |||
| GenBank mRNA | BC137523.1 | - | 1 | 277 | 9021523 | |
| BC137524.1 | - | 1 | 277 | 9021524 | ||

OR5H6
Genomic Map