siRNA Details
| Gene Symbol: OR4K5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005483.1 | NP_001005483.1 | 1 | 650 | ||
| GenBank mRNA | BC140947.1 | - | 1 | 675 | 9056759 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4K5| Gene Symbol: OR4K5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005483.1 | NP_001005483.1 | 1 | 650 | ||
| GenBank mRNA | BC140947.1 | - | 1 | 675 | 9056759 | |