siRNA Details
| Gene Symbol: OR51G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005237.1 | NP_001005237.1 | 1 | 59 | ||
| GenBank mRNA | BC136852.1 | - | 1 | 127 | 9020843 | |
| BC136861.1 | - | 1 | 127 | 9020852 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR51G1| Gene Symbol: OR51G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005237.1 | NP_001005237.1 | 1 | 59 | ||
| GenBank mRNA | BC136852.1 | - | 1 | 127 | 9020843 | |
| BC136861.1 | - | 1 | 127 | 9020852 | ||