siRNA Details
Gene Symbol: CRYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001888.4 | NP_001879.1 | 5 | 694 | ||
GenBank mRNA | AK290852.1 | - | 3 | 413 | ||
AK310424.1 | - | 3 | 367 | |||
BC018061.1 | - | 3 | 412 | 4817634 | ||
BI860684.1 | - | 4 | 367 | |||
BX648477.1 | - | 5 | 610 | |||
L02950.1 | - | 3 | 373 | |||
U85772.1 | - | 3 | 368 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016669, AF039391, AK143116, AK138422, BC045159, and others.
Panther Classification:
Gene Ontology Categories:
