siRNA Details
| Gene Symbol: FCRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159488.1 | NP_001152960.1 | 7 | 1238 | ||
| NM_030764.3 | NP_110391.2 | 7 | 1238 | |||
| NR_125358.1 | 4 | 386 | ||||
| XM_011509974.2 | XP_011508276.1 | 7 | 1573 | |||
| XM_011509975.2 | XP_011508277.1 | 7 | 1575 | |||
| XM_011509976.2 | XP_011508278.1 | 7 | 1574 | |||
| XM_017002316.1 | XP_016857805.1 | 7 | 1573 | |||
| XM_017002317.1 | XP_016857806.1 | 7 | 1376 | |||
| XM_017002318.1 | XP_016857807.1 | 6 | 1296 | |||
| XM_017002319.1 | XP_016857808.1 | 6 | 1288 | |||
| GenBank mRNA | AF319438.1 | - | 2 | 723 | ||
| AF319439.1 | - | 2 | 561 | |||
| AF319440.1 | - | 1 | 970 | |||
| AF390037.1 | - | 2 | 561 | |||
| AF459633.1 | - | 7 | 1226 | |||
| AK301113.1 | - | 7 | 1225 | |||
| AL833194.1 | - | 2 | 3097 | |||
| AY043465.1 | - | 7 | 1225 | |||
| AY358130.1 | - | 7 | 1228 | |||
| BC069185.1 | - | 4 | 390 | |||
| BC129836.1 | - | 2 | 563 | 40115446 | ||

FCRL2
Genomic Map