siRNA Details
| Gene Symbol: FAM65A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193522.1 | NP_001180451.1 | 14 | 2694 | ||
| NM_001193523.1 | NP_001180452.1 | 14 | 2807 | |||
| NM_001193524.1 | NP_001180453.1 | 14 | 2720 | |||
| NM_024519.3 | NP_078795.2 | 14 | 2685 | |||
| XM_011523321.1 | XP_011521623.1 | 14 | 3499 | |||
| XM_011523322.1 | XP_011521624.1 | 14 | 2694 | |||
| XM_011523324.2 | XP_011521626.1 | 14 | 2754 | |||
| XM_011523325.1 | XP_011521627.1 | 14 | 2709 | |||
| XM_011523326.1 | XP_011521628.1 | 14 | 2908 | |||
| XM_017023663.1 | XP_016879152.1 | 14 | 2691 | |||
| XM_017023664.1 | XP_016879153.1 | 13 | 2580 | |||
| GenBank mRNA | AB067517.1 | - | 13 | 2540 | ||
| AK023787.1 | - | 9 | 2177 | |||
| AK127792.1 | - | 14 | 2694 | |||
| AK293748.1 | - | 14 | 2717 | |||
| AK295677.1 | - | 14 | 2651 | |||
| AK302868.1 | - | 4 | 1841 | |||
| AL834312.1 | - | 4 | 1750 | |||
| BC054512.1 | - | 4 | 1727 | |||
| BC098587.1 | - | 8 | 2122 | |||

FAM65A
Genomic Map