siRNA Details
| Gene Symbol: C2orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024520.2 | NP_078796.2 | 4 | 908 | ||
| XM_005246852.2 | XP_005246909.1 | 3 | 819 | |||
| GenBank mRNA | AK026208.1 | - | 4 | 908 | ||
| AL832964.1 | - | 3 | 227 | |||
| BC017959.1 | - | 3 | 592 | 4607945 | ||

C2orf47
Genomic Map