siRNA Details
| Gene Symbol: C2orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024520.2 | NP_078796.2 | 5 | 983 | ||
| GenBank mRNA | AK026208.1 | - | 5 | 983 | ||
| AL832964.1 | - | 4 | 302 | |||
| BC017959.1 | - | 4 | 667 | 4607945 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf47| Gene Symbol: C2orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024520.2 | NP_078796.2 | 5 | 983 | ||
| GenBank mRNA | AK026208.1 | - | 5 | 983 | ||
| AL832964.1 | - | 4 | 302 | |||
| BC017959.1 | - | 4 | 667 | 4607945 | ||