siRNA Details
| Gene Symbol: ATP13A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024524.3 | NP_078800.3 | 29 | 3578 | ||
| XM_005269357.3 | XP_005269414.1 | 29 | 3599 | |||
| XM_011513120.2 | XP_011511422.1 | 30 | 3700 | |||
| XM_011513121.1 | XP_011511423.1 | 29 | 3578 | |||
| XM_011513122.1 | XP_011511424.1 | 29 | 3272 | |||
| XM_011513123.2 | XP_011511425.1 | 30 | 3705 | |||
| XM_011513124.2 | XP_011511426.1 | 30 | 3705 | |||
| GenBank mRNA | AK024639.1 | - | 18 | 1975 | ||
| AK075094.1 | - | 10 | 1023 | |||
| AK225064.1 | - | 18 | 1975 | |||

ATP13A3
Genomic Map