siRNA Details
| Gene Symbol: CEP97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303401.1 | NP_001290330.1 | 2, 3 | 262 | ||
| NM_024548.3 | NP_078824.2 | 2, 3 | 262 | |||
| XM_006713743.3 | XP_006713806.1 | 2, 3 | 755 | |||
| XM_011513127.1 | XP_011511429.1 | 2, 3 | 197 | |||
| XM_017007178.1 | XP_016862667.1 | 2, 3 | 757 | |||
| GenBank mRNA | AK309806.1 | - | 2, 3 | 263 | ||
| AL833269.1 | - | 2, 3 | 212 | |||
| BC041085.1 | - | 2, 3 | 205 | 5528397 | ||
| DA520309.1 | - | 2, 3 | 264 | |||

CEP97
Genomic Map