siRNA Details
| Gene Symbol: ADIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024551.2 | NP_078827.2 | 2 | 256 | ||
| XM_005253789.1 | XP_005253846.1 | 2 | 378 | |||
| XM_006719018.2 | XP_006719081.1 | 3 | 375 | |||
| XM_011521024.2 | XP_011519326.1 | 3 | 386 | |||
| GenBank mRNA | AK127196.1 | - | 3 | 311 | ||
| AK128511.1 | - | 2 | 310 | |||
| AY424280.1 | - | 1 | 135 | |||
| BC051858.2 | - | 2 | 256 | 6527208 | ||

ADIPOR2
Genomic Map