siRNA Details
| Gene Symbol: TANGO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024562.1 | NP_078838.1 | 2 | 422 | ||
| XM_011523327.2 | XP_011521629.1 | 2 | 481 | |||
| GenBank mRNA | AB051533.1 | - | 2 | 422 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TANGO6| Gene Symbol: TANGO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024562.1 | NP_078838.1 | 2 | 422 | ||
| XM_011523327.2 | XP_011521629.1 | 2 | 481 | |||
| GenBank mRNA | AB051533.1 | - | 2 | 422 | ||