siRNA Details
| Gene Symbol: ARMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024573.2 | NP_078849.1 | 2 | 377 | ||
| GenBank mRNA | AK022972.1 | - | 2 | 348 | ||
| AL110241.1 | - | 2 | 103 | |||
| BC011348.1 | - | 2 | 97 | 4340090 | ||
| DB443862.1 | - | 2 | 377 | |||

ARMT1
Genomic Map