siRNA Details
| Gene Symbol: C1orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301039.1 | NP_001287968.1 | 3 | 253 | ||
| NM_001301040.1 | NP_001287969.1 | 3 | 253 | |||
| NM_001301042.1 | NP_001287971.1 | 2 | 123 | |||
| NM_024579.3 | NP_078855.2 | 2 | 142 | |||
| GenBank mRNA | AA095731.1 | - | 2 | 110 | ||
| AK026874.1 | - | 2 | 127 | |||
| AX770997.1 | - | 2 | 140 | |||
| AX770998.1 | - | 3 | 167 | |||
| AX770999.1 | - | 2 | 142 | |||
| BC017761.1 | - | 2 | 127 | 4617936 | ||
| T82829.1 | - | 3 | 233 | |||

C1orf54
Genomic Map