siRNA Details
| Gene Symbol: SCRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193528.1 | NP_001180457.1 | 4 | 455 | ||
| NM_024583.4 | NP_078859.2 | 4 | 546 | |||
| XM_005246853.2 | XP_005246910.1 | 4 | 951 | |||
| XM_005246854.4 | XP_005246911.1 | 4 | 2111 | |||
| XM_005246855.2 | XP_005246912.1 | 4 | 999 | |||
| XM_005246856.2 | XP_005246913.1 | 4 | 955 | |||
| XM_005246857.2 | XP_005246914.1 | 4 | 655 | |||
| XM_011511839.2 | XP_011510141.1 | 3 | 355 | |||
| XM_017004909.1 | XP_016860398.1 | 4 | 1543 | |||
| XM_017004910.1 | XP_016860399.1 | 4 | 1021 | |||
| GenBank mRNA | AK057912.1 | - | 4 | 524 | ||
| AK295357.1 | - | 4 | 439 | |||
| BC119684.1 | - | 4 | 497 | 40116568 | ||
| BC119685.1 | - | 4 | 497 | 40116570 | ||
| BM725063.1 | - | 3 | 537 | |||

SCRN3
Genomic Map