siRNA Details
| Gene Symbol: ARMC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304271.1 | NP_001291200.1 | 3 | 1260 | ||
| NM_024585.3 | NP_078861.1 | 3 | 1324 | |||
| GenBank mRNA | AK025813.1 | - | 3 | 1132 | ||
| AK309353.1 | - | 2 | 1099 | |||
| BC011728.1 | - | 3 | 1289 | 4423125 | ||

ARMC7
Genomic Map