siRNA Details
| Gene Symbol: TMEM53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024587.3 | NP_078863.2 | 2, 3 | 410 | ||
| GenBank mRNA | AK026006.1 | - | 2, 3 | 217 | ||
| AK225405.1 | - | 2, 3 | 217 | |||
| BC064520.1 | - | 2, 3 | 229 | 5582377 | ||
| CR457348.1 | - | 2, 3 | 193 | |||
| HY108001.1 | - | 2, 3 | 412 | |||

TMEM53
Genomic Map