siRNA Details
| Gene Symbol: SRD5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024592.4 | NP_078868.1 | 5 | 957 | ||
| XM_005265766.3 | XP_005265823.1 | 4 | 929 | |||
| XM_005265767.3 | XP_005265824.1 | 3 | 617 | |||
| XM_017008601.1 | XP_016864090.1 | 5 | 732 | |||
| GenBank mRNA | AK023414.1 | - | 5 | 937 | ||
| BC002480.2 | - | 5 | 933 | 3350037 | ||
| CR457312.1 | - | 5 | 841 | |||

SRD5A3
Genomic Map