siRNA Details
| Gene Symbol: SRD5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024592.4 | NP_078868.1 | 4 | 797 | ||
| XM_017008601.1 | XP_016864090.1 | 4 | 572 | |||
| GenBank mRNA | AK023414.1 | - | 4 | 777 | ||
| BC002480.2 | - | 4 | 773 | 3350037 | ||
| CR457312.1 | - | 4 | 681 | |||

SRD5A3
Genomic Map