siRNA Details
| Gene Symbol: PANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024594.3 | NP_078870.1 | 2 | 407 | ||
| GenBank mRNA | AK022961.1 | - | 2 | 152 | ||
| AK223285.1 | - | 2 | 136 | |||
| BC013705.1 | - | 2 | 381 | 4343270 | ||
| CB045475.1 | - | 2 | 404 | |||

PANK3
Genomic Map