siRNA Details
| Gene Symbol: RPAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146075.1 | NP_001139547.1 | 14 | 1790 | ||
| NM_001146076.1 | NP_001139548.1 | 14 | 1733 | |||
| NM_024604.2 | NP_078880.2 | 15 | 1892 | |||
| XM_011538741.2 | XP_011537043.1 | 13 | 1643 | |||
| GenBank mRNA | AK025561.1 | - | 15 | 1822 | ||
| AK299465.1 | - | 14 | 1698 | |||
| BC056415.1 | - | 14 | 1715 | 5218249 | ||

RPAP3
Genomic Map