siRNA Details
| Gene Symbol: CSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000759.3 | NP_000750.1 | 5 | 937 | ||
| NM_001178147.1 | NP_001171618.1 | 4 | 820 | |||
| NM_172219.2 | NP_757373.1 | 5 | 928 | |||
| NM_172220.2 | NP_757374.2 | 4 | 829 | |||
| NR_033662.1 | 4 | 1104 | ||||
| GenBank mRNA | AK298116.1 | - | 3 | 980 | ||
| BC033245.1 | - | 4 | 998 | 5434958 | ||
| X03438.1 | - | 5 | 854 | |||
| X03655.1 | - | 5 | 844 | |||

CSF3
Genomic Map