siRNA Details
| Gene Symbol: VEPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167911.1 | NP_001161383.1 | 9 | 1846 | ||
| NM_001167912.1 | NP_001161384.1 | 9 | 1846 | |||
| NM_024621.2 | NP_078897.2 | 9 | 2037 | |||
| XM_011513134.2 | XP_011511436.1 | 8 | 1780 | |||
| XM_011513135.1 | XP_011511437.1 | 9 | 1676 | |||
| GenBank mRNA | AB051479.1 | - | 9 | 1846 | ||
| AK027625.1 | - | 9 | 1935 | |||
| AL713656.1 | - | 8 | 1634 | |||
| BC026235.1 | - | 1 | 159 | |||
| BC042159.1 | - | 2 | 496 | |||
| BC101660.1 | - | 8 | 1673 | 8069166 | ||
| BC111017.1 | - | 9 | 1838 | 5174889 | ||
| BC113555.1 | - | 8 | 1673 | 8322607 | ||

VEPH1
Genomic Map