siRNA Details
| Gene Symbol: C22orf29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024627.5 | NP_078903.3 | 3 | 1068 | ||
| GenBank mRNA | AK024778.1 | - | 1 | 444 | ||
| AK225088.1 | - | 1 | 444 | |||
| AK291262.1 | - | 3 | 1036 | |||
| AL832509.1 | - | 3 | 957 | |||
| BC011679.2 | - | 1 | 575 | 4303449 | ||
| BX640785.1 | - | 2 | 1157 | |||
| BX640969.1 | - | 3 | 1037 | |||
| BX640998.1 | - | 3 | 1037 | |||

C22orf29
Genomic Map