siRNA Details
| Gene Symbol: QTRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256835.1 | NP_001243764.1 | 4 | 470 | ||
| NM_024638.3 | NP_078914.1 | 5 | 578 | |||
| XM_005247769.3 | XP_005247826.1 | 5 | 359 | |||
| XM_006713744.3 | XP_006713807.1 | 4 | 211 | |||
| XM_011513137.2 | XP_011511439.1 | 4 | 441 | |||
| GenBank mRNA | AK023022.1 | - | 5 | 468 | ||
| AL833709.1 | - | 4 | 438 | |||
| BC034559.2 | - | 4 | 452 | 4385906 | ||
| BC039265.1 | - | 5 | 549 | 5296335 | ||
| CR457296.1 | - | 3 | 319 | |||
| DC400836.1 | - | 4 | 470 | |||

QTRT2
Genomic Map