siRNA Details
| Gene Symbol: ZNF322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242797.1 | NP_001229726.1 | 5 | 877 | ||
| NM_001242798.1 | NP_001229727.1 | 3 | 732 | |||
| NM_001242799.1 | NP_001229728.1 | 3 | 713 | |||
| NM_024639.4 | NP_078915.2 | 4 | 802 | |||
| GenBank mRNA | AK027046.1 | - | 1 | 122 | ||
| AK290038.1 | - | 4 | 527 | |||
| AY376736.1 | - | 4 | 585 | |||
| BC050425.1 | - | 1 | 298 | 6473688 | ||
| BC074772.2 | - | 1 | 94 | |||
| BC119669.2 | - | 1 | 215 | 40116184 | ||

ZNF322
Genomic Map