siRNA Details
| Gene Symbol: ZMAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135731.1 | NP_001129203.1 | 2, 3 | 247 | ||
| NM_024645.2 | NP_078921.1 | 2, 3 | 247 | |||
| XM_017013836.1 | XP_016869325.1 | 2, 3 | 251 | |||
| GenBank mRNA | AK023904.1 | - | 2, 3 | 247 | ||
| BC019598.1 | - | 2, 3 | 233 | 4934088 | ||

ZMAT4
Genomic Map