siRNA Details
| Gene Symbol: C11orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302084.1 | NP_001289013.1 | 8 | 896 | ||
| NM_024650.3 | NP_078926.3 | 10 | 1185 | |||
| GenBank mRNA | AK026184.1 | - | 9 | 986 | ||
| AK225435.1 | - | 9 | 986 | |||
| AK302025.1 | - | 8 | 848 | |||
| BC028240.1 | - | 10 | 1188 | 5243907 | ||

C11orf80
Genomic Map