siRNA Details
| Gene Symbol: CSN1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025104.1 | NP_001020275.1 | 4, 5 | 155 | ||
| NM_001890.1 | NP_001881.1 | 4, 5 | 155 | |||
| XM_006714089.2 | XP_006714152.1 | 4, 5 | 174 | |||
| XM_006714090.2 | XP_006714153.1 | 4, 5 | 174 | |||
| XM_006714091.2 | XP_006714154.1 | 4, 5 | 174 | |||
| GenBank mRNA | BC128227.1 | - | 4, 5 | 144 | 40114618 | |
| BC128228.1 | - | 4, 5 | 144 | 40114622 | ||
| BG217918.1 | - | 3, 4 | 117 | |||
| DQ064604.1 | - | 3, 4 | 106 | |||
| U23157.1 | - | 4, 5 | 134 | |||
| X78416.1 | - | 4, 5 | 155 | |||
| X98084.1 | - | 3, 4 | 83 | |||

CSN1S1
Genomic Map